Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Timp
- Entrez Gene ID
-
41248
- Gene Name
- Tissue inhibitor of metalloproteases
- Gene Aliases
-
CG6281, DTIMP, Dmel\CG6281, Dmel_CG6281, Dtimp, TIMP, TIMP1, dTIMP, timp
- Chromosome Location
-
Chr.3R: 10203875 - 10213007 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0025879
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169336.4 |
NP_731461.1 |
2 - 3 |
456 |
73 |
GenBank mRNA |
AY069211.1 |
- |
2 - 3 |
437 |
73 |
View Full Product Details