Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or92a
- Entrez Gene ID
-
42425
- Gene Name
- Odorant receptor 92a
- Gene Aliases
-
92E.1, 92a, CG17916, DOR111, Dmel\CG17916, Dmel_CG17916, OR 92a, Or92A
- Chromosome Location
-
Chr.3R: 20504788 - 20506186 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038798
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079690.2 |
NP_524414.2 |
2 - 3 |
1018 |
66 |
View Full Product Details