Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi1
Entrez Gene ID
40752
Gene Name
Osiris 1
Gene Aliases
CG15585, Dmel\CG15585, Dmel_CG15585, osi1
Chromosome Location
Chr.3R: 6173549 - 6174971 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037406
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141361.2 NP_649618.1 1 - 2 488 76
GenBank mRNA BT100152.4 - 1 - 2 392 76

View Full Product Details