Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Npc2c
- Entrez Gene ID
-
41233
- Gene Name
- Niemann-Pick type C-2c
- Gene Aliases
-
CG3934, DmML5A, Dmel\CG3934, Dmel_CG3934, npc2c
- Chromosome Location
-
Chr.3R: 10060252 - 10061136 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037783
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141719.2 |
NP_649976.1 |
1 - 2 |
134 |
72 |
View Full Product Details