Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beat-Va
Entrez Gene ID
41578
Gene Name
beaten path Va
Gene Aliases
Beat-Va, CG10134, Dmel\CG10134, Dmel_CG10134, beat Va
Chromosome Location
Chr.3R: 12840560 - 12848419 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038087
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202285.1 NP_001189214.1 4 - 5 1045 57
NM_001275581.1 NP_001262510.1 4 - 5 1045 57
NM_141976.4 NP_650233.3 4 - 5 1045 57
GenBank mRNA BT044588.1 - 4 - 5 1046 57

View Full Product Details