Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kal1
Entrez Gene ID
42865
Gene Name
Kallmann syndrome 1
Gene Aliases
CG6173, CT19368, DmKal-1, Dmel\CG6173, Dmel_CG6173, Dmkal, Kal-1, kal-1
Chromosome Location
Chr.3R: 24141395 - 24147490 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039155
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142963.3 NP_651220.3 6 - 7 1702 65
GenBank mRNA AF342988.1 - 6 - 7 1665 65
BT015965.1 - 6 - 7 1703 65
BT044162.1 - 6 - 7 1657 65

View Full Product Details