Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ymp
Entrez Gene ID
43088
Gene Name
yellow-emperor
Gene Aliases
BcDNA:AT03213, CG12250, Dmel\CG12250, Dmel_CG12250
Chromosome Location
Chr.3R: 25591502 - 25609127 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261287
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300601.1 NP_001287530.1 2 - 3 206 88
NM_001300602.1 NP_001287531.1 2 - 3 206 88
NM_143147.3 NP_651404.1 2 - 3 206 88
NM_170230.2 NP_733109.1 2 - 3 206 88
GenBank mRNA AY118672.1 - 2 - 3 197 88
KX531975.1 - 2 - 3 110 88

View Full Product Details