Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk15
Entrez Gene ID
43188
Gene Name
pickpocket 15
Gene Aliases
CG14239, Dmel\CG14239, Dmel_CG14239
Chromosome Location
Chr.3R: 26401909 - 26403656 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039424
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104467.1 NP_001097937.1 4 - 5 1226 74

View Full Product Details