Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpr97Ea
Entrez Gene ID
43258
Gene Name
Cuticular protein 97Ea
Gene Aliases
CG6131, Dmel\CG6131, Dmel_CG6131
Chromosome Location
Chr.3R: 27083130 - 27087778 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039480
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276077.1 NP_001263006.1 3 - 4 729 136
NM_143272.2 NP_651529.2 3 - 4 741 136
GenBank mRNA BT150156.1 - 3 - 4 741 136

View Full Product Details