Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eater
- Entrez Gene ID
-
43262
- Gene Name
- CG6124-PC
- Gene Aliases
-
C901-like, CG6124, CT19227, Dmel\CG6124, Dmel_CG6124, Eater
- Chromosome Location
-
Chr.3R: 27095867 - 27099679 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0243514
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143276.3 |
NP_651533.3 |
1 - 2 |
118 |
77 |
View Full Product Details