Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bigmax
- Entrez Gene ID
-
43293
- Gene Name
- CG3350-PA
- Gene Aliases
-
BIGMAX, Bigmax, CG3350, DmMlx, Dmel\CG3350, Dmel_CG3350, Mlx, bHLHd13, cg3350, mlx
- Chromosome Location
-
Chr.3R: 27287366 - 27288526 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039509
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143299.3 |
NP_651556.2 |
2 - 3 |
605 |
93 |
View Full Product Details