Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tusp
Entrez Gene ID
43317
Gene Name
CG5586-PD
Gene Aliases
CG5586, DMG2-MB6.chr3R.24.043, Dmel\CG5586, Dmel_CG5586, TUSP
Chromosome Location
Chr.3R: 27607879 - 27634932 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039530
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202374.2 NP_001189303.1 5 - 6 2038 88
NM_001276085.2 NP_001263014.1 5 - 6 1672 88
NM_001276086.2 NP_001263015.1 5 - 6 1672 88
NM_143316.4 NP_651573.4 5 - 6 1438 88
GenBank mRNA BT132902.1 - 5 - 6 1438 88

View Full Product Details