Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CheB98a
- Entrez Gene ID
-
50092
- Gene Name
- Chemosensory protein A 98a
- Gene Aliases
-
CG14531, CheB98ap, Dmel\CG14531, Dmel_CG14531
- Chromosome Location
-
Chr.3R: 28806702 - 28807501 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261289
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144012.3 |
NP_652269.3 |
1 - 1 |
241 |
56 |
View Full Product Details