Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CG46339
Entrez Gene ID
44359
Gene Name
CG46339-PA
Gene Aliases
BcDNA:GH07466, CG5518, Dmel\CG46339, Dmel_CG46339, sda
Chromosome Location
Chr.3R: 26915761 - 26945309 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285963
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104469.3 NP_001097939.1 12 - 13 4109 61
NM_001260394.2 NP_001247323.1 12 - 13 3812 61
NM_001276072.1 NP_001263001.1 13 - 14 4145 61
NM_001363935.1 NP_001350863.1 10 - 11 3680 61
NM_001363936.1 NP_001350864.1 11 - 12 4013 61
NM_144487.3 NP_652744.2 11 - 12 3776 61
GenBank mRNA AF480087.1 - 11 - 12 3753 61
AY122083.1 - 11 - 12 3816 61

View Full Product Details