Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nmdar1
Entrez Gene ID
40665
Gene Name
NMDA receptor 1
Gene Aliases
CG2902, DMNMDAR1, DNMDAR-I, DNmdar, DmelNmdar1, Dmel\CG2902, Dmel_CG2902, DrNR1, NMDA, NMDA R1, NMDA-R1, NMDAR, NMDAR1, NMdar1, NR1, NRI, NmdaR1, Nmdar, dNR1, dnmdar-I, dsNR1, nmdar1, nmr, nr1
Chromosome Location
Chr.3R: 5508842 - 5523328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010399
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169059.2 NP_730940.1 11 - 12 2028 64
GenBank mRNA AY070577.1 - 11 - 12 2024 64
X71790.1 - - 1936 64

View Full Product Details