Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plx
Entrez Gene ID
40704
Gene Name
pollux
Gene Aliases
CG1093, CG14674, CG34122, CG42612, Dmel\CG42612, Dmel_CG42612, Plx
Chromosome Location
Chr.3R: 5685855 - 5712837 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261261
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170040.2 NP_001163511.1 7 - 8 3157 62
NM_001275368.1 NP_001262297.1 6 - 7 3247 62
NM_001275369.1 NP_001262298.1 4 - 5 2142 62
NM_169094.2 NP_730988.1 4 - 5 2397 62
NM_169095.3 NP_730989.2 5 - 6 2304 62
GenBank mRNA BT133115.1 - 7 - 8 2880 62

View Full Product Details