Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mhcl
Entrez Gene ID
41955
Gene Name
Myosin heavy chain-like
Gene Aliases
CG10218, CG10224, CG31045, DmPDZ, Dmel\CG31045, Dmel_CG31045, MHCL(PDZ), Myo7A
Chromosome Location
Chr.3R: 16068523 - 16098761 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026059
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043250.2 NP_001036715.1 14 - 15 5078 77
NM_001043251.2 NP_001036716.1 11 - 12 4805 77
NM_001275694.1 NP_001262623.1 4 - 5 2333 77
NM_169700.3 NP_732108.2 14 - 15 5078 77
NM_169701.3 NP_732109.2 14 - 15 5078 77
NM_169703.1 NP_732111.1 3 - 4 2396 77
NM_170650.4 NP_732110.2 5 - 6 1219 77
GenBank mRNA BT001428.1 - 5 - 6 1219 77

View Full Product Details