Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SNF4Agamma
- Entrez Gene ID
-
42515
- Gene Name
- SNF4/AMP-activated protein kinase gamma subunit
- Gene Aliases
-
0050/42, 0127/19, AMPK, AMPK&ggr, AMPK-&ggr, AMPKgamma, AMPgamma, CG17299, CG5806, Dmel\CG17299, Dmel_CG17299, EP3015b, FBgn0025803, SNF4, SNF4A, SNF4A-gamma, SNF4Ag, SNF4a, SNF4gamma, Snfg, Snfgamma, anon-WO0118547.338, anon-WO0118547.575, anon-WO0257455.21, l(3)S005042, l(3)S012719, loe, loeI, snf4
- Chromosome Location
-
Chr.3R: 21140739 - 21214269 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264357
View Full Product Details