Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cad96Cb
- Entrez Gene ID
-
43033
- Gene Name
- Cadherin 96Cb
- Gene Aliases
-
CG10421, CG13664, CT29262, Cad96C, DCad96Cb, Dmel\CG13664, Dmel_CG13664, cad96C, cad96Cb, cg13664
- Chromosome Location
-
Chr.3R: 25224156 - 25232321 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039294
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143098.4 |
NP_651355.3 |
4 - 5 |
1840 |
65 |
View Full Product Details