Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp96F
Entrez Gene ID
43127
Gene Name
Tetraspanin 96F
Gene Aliases
BcDNA:LD19727, CG6120, Dm.Tsp96F, Dmel\CG6120, Dmel_CG6120, LD19727
Chromosome Location
Chr.3R: 25872900 - 25881427 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027865
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276047.2 NP_001262976.1 4 - 5 1004 74
NM_001276048.2 NP_001262977.1 4 - 5 1004 74
NM_079776.3 NP_524500.1 4 - 5 1004 74
GenBank mRNA AF160941.1 - 4 - 5 951 74

View Full Product Details