Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DNApol-alpha73
- Entrez Gene ID
-
43278
- Gene Name
- DNA polymerase alpha 73kD subunit
- Gene Aliases
-
CG5923, DNA pol-73, DNA polalpha, DNA polymerase alpha 73 kD, DNApol-alpha, DNApol-alpha72, DNApolalpha, Dmel\CG5923, Dmel_CG5923, Dmpol alpha, POLA, Pol, Polalpha, Polalpha 73KDa, Polalpha 73kDa, Polalpha73, pol alpha, polalpha
- Chromosome Location
-
Chr.3R: 27239163 - 27241228 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0005696
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170327.4 |
NP_733206.3 |
1 - 1 |
1898 |
76 |
View Full Product Details