Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhc98D
Entrez Gene ID
43379
Gene Name
Dynein heavy chain at 89D
Gene Aliases
CG1842, DHC, DHC6, Dhc, Dmel\CG1842, Dmel_CG1842, dhc
Chromosome Location
Chr.3R: 28486953 - 28519398 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013813
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170288.1 NP_001163759.1 18 - 19 6065 98
NM_001276107.1 NP_001263036.1 12 - 13 3747 98
NM_079817.3 NP_524541.3 19 - 20 6086 98

View Full Product Details