Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chp
Entrez Gene ID
43690
Gene Name
chaoptin
Gene Aliases
24B10, 516, BSH, CG1744, CHP, CT5040, Chaoptin, Chp, Dmel\CG1744, Dmel_CG1744, MAb 24B10, MAb24B10, Mab24B10, Nsp, idl-A, mAB24B10, mAb 24B10, mAb24B10
Chromosome Location
Chr.3R: 31204180 - 31210730 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267435
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276205.1 NP_001263134.1 14 - 15 4293 58
NM_001276206.1 NP_001263135.1 14 - 15 4305 58
NM_001276207.1 NP_001263136.1 14 - 15 4284 58
NM_001276208.1 NP_001263137.1 14 - 15 4305 58
NM_079866.3 NP_524605.1 14 - 15 4314 58
GenBank mRNA BT126010.1 - 14 - 15 4092 58

View Full Product Details