Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IKKbeta
- Entrez Gene ID
-
44432
- Gene Name
- I-kappaB kinase beta
- Gene Aliases
-
CG4201, DIK, DLAK, DmIKK, DmIKKBeta, DmIKKb, DmIKKbeta, DmIkkbeta, Dmel\CG4201, Dmel_CG4201, Dmikkb, Dmikkbeta, IK, IKK, IKK-beta, IKKB, IKKb, IRD-5, IRD5, Ik, Ikkb, Ikkbeta, Ird-5, Ird5, LRK, Lak, anon-89Bd, dIKK, dIKK-beta, dIKKbeta, dLak, dmIKKbeta, ikk, ird, ird-5, ird5, ird5/DmIkk[beta], ird[5]
- Chromosome Location
-
Chr.3R: 16046347 - 16049421 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024222
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080012.4 |
NP_524751.3 |
1 - 2 |
205 |
85 |
View Full Product Details