Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pum
- Entrez Gene ID
-
41094
- Gene Name
- pumilio
- Gene Aliases
-
CG9755, CG9763, Dmel\CG9755, Dmel_CG9755, EP(3)0883, PKL, PUM, Pum, anon-WO0172774.19, bem, fs(3)02003, l(3)01688, ova, ovt, pkl
- Chromosome Location
-
Chr.3R: 9066343 - 9237682 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003165
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176427.3 |
NP_788604.1 |
2 - 3 |
871 |
59 |
GenBank mRNA |
BT001872.1 |
- |
2 - 3 |
271 |
59 |
View Full Product Details