Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Synd
Entrez Gene ID
42467
Gene Name
Syndapin
Gene Aliases
CG15694, CG33094, D-Synd, Dmel\CG33094, Dmel_CG33094, synd
Chromosome Location
Chr.3R: 20832799 - 20840118 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053094
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275857.1 NP_001262786.1 8 - 9 1292 139
NM_001275858.1 NP_001262787.1 8 - 9 1298 139
NM_176520.2 NP_788697.1 8 - 9 1295 139
GenBank mRNA AY069690.1 - 8 - 9 1268 139

View Full Product Details