Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tpi
- Entrez Gene ID
-
43582
- Gene Name
- Triose phosphate isomerase
- Gene Aliases
-
CG2171, Dmel\CG2171, Dmel_CG2171, ND14, TPI, TPIS, sgk, tpi, wstd
- Chromosome Location
-
Chr.3R: 30132981 - 30134599 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086355
- Gene Symbol
- CG1607
- Entrez Gene ID
-
43707
- Gene Name
- CG1607-PA
- Gene Aliases
-
Cg1607, Dmel\CG1607, Dmel_CG1607
- Chromosome Location
-
Chr.3R: 31382109 - 31391893 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039844
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT029261.1 |
- |
- |
5062 |
109 |
View Full Product Details