Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AttA
- Entrez Gene ID
-
36636
- Gene Name
- Attacin-A
- Gene Aliases
-
AHA, ATT, Att, Att A, Att-A, BcDNA:LP05763, CG10146, Dmel\CG10146, Dmel_CG10146, anon-WO0140519.5, att, att A, attA
- Chromosome Location
-
Chr.2R: 14747362 - 14748215 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0012042
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079021.5 |
NP_523745.1 |
2 - 2 |
662 |
68 |
View Full Product Details