Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp6a9
- Entrez Gene ID
-
36663
- Gene Name
- Cytochrome P450-6a9
- Gene Aliases
-
6a9, CG10246, CYP6A9, CYP6a9, Cyp6A9, Cyt-P450-rAF4, Dmel\CG10246, Dmel_CG10246, P-450, P450
- Chromosome Location
-
Chr.2R: 14879356 - 14881372 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013771
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079024.3 |
NP_523748.2 |
2 - 2 |
1551 |
152 |
View Full Product Details