Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubi-p63E
Entrez Gene ID
38456
Gene Name
Ubiquitin-63E
Gene Aliases
CG11624, DmUb, DmUbi-p63E, Dmel\CG11624, Dmel_CG11624, UB, Ub, Ub-p63E, Ubi-P63e, Ubi-p, Ubi63E, Ubi63p, Ubip, Ubip63E, Ubiq, Ubp, magn, ubi, ubi-p63E, ubi63E
Chromosome Location
Chr.3L: 3899259 - 3903184 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003943
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274454.1 NP_001261383.1 2 - 2 2487 85
NM_079185.4 NP_523909.2 3 - 3 2622 85
NM_206272.1 NP_995994.1 2 - 2 2573 85

View Full Product Details