Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ccp84Ad
- Entrez Gene ID
-
40822
- Gene Name
- CG2341-PA
- Gene Aliases
-
BG:DS00004.4, CG2341, Ccp, DmelCcp84Ad, Dmel\CG2341, Dmel_CG2341, cc5
- Chromosome Location
-
Chr.3R: 6692732 - 6693527 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004780
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141423.4 |
NP_649680.1 |
2 - 2 |
332 |
124 |
View Full Product Details