Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pHCl-1
Entrez Gene ID
39730
Gene Name
pH-sensitive chloride channel 1
Gene Aliases
BP1024, BcDNA:GH21757, CG12486, CG13444, CG33989, CG44099, CG6112, CT19189, Dmel\CG44099, Dmel_CG44099, pHCL-A, pHCl
Chromosome Location
Chr.3L: 15848578 - 15899029 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038935.3 NP_001034024.2 6 - 7 1595 82
NM_001038936.4 NP_001034025.2 6 - 7 1592 82
NM_001038937.2 NP_001034026.1 6 - 7 1595 82
NM_001169979.1 NP_001163450.1 6 - 7 1595 82
NM_001259852.1 NP_001246781.1 6 - 7 1600 82
NM_001274965.1 NP_001261894.1 5 - 6 1579 82
NM_001274968.1 NP_001261897.1 5 - 6 1574 82
NM_001274969.1 NP_001261898.1 6 - 7 1595 82
NM_001274971.1 NP_001261900.1 5 - 6 1574 82
NM_001370024.1 NP_001356981.1 5 - 6 1579 82
NM_001370025.1 NP_001356910.1 6 - 7 1600 82
GenBank mRNA BT012472.1 - 6 - 7 1596 82

View Full Product Details