Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pHCl-1
Entrez Gene ID
39730
Gene Name
pH-sensitive chloride channel 1
Gene Aliases
BP1024, BcDNA:GH21757, CG12486, CG13444, CG33989, CG44099, CG6112, CT19189, Dmel\CG44099, Dmel_CG44099, pHCL-A, pHCl
Chromosome Location
Chr.3L: 15848578 - 15899029 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038935.3 NP_001034024.2 12 - 13 2328 69
NM_001038936.4 NP_001034025.2 12 - 13 2325 69
NM_001169979.1 NP_001163450.1 12 - 13 2328 69
NM_001259853.1 NP_001246782.1 11 - 12 2258 69
NM_001274966.1 NP_001261895.1 10 - 11 2237 69
NM_001274970.1 NP_001261899.1 10 - 11 2232 69
NM_001370025.1 NP_001356910.1 12 - 13 2333 69

View Full Product Details