Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nvd
Entrez Gene ID
5740633
Gene Name
neverland
Gene Aliases
80Fc, CG40050, Dmel\CG40050, Dmel_CG40050, Nvd, l(3)80Fc, l(3)Lh6, l6, nvd-Dm
Chromosome Location
Chr.3L: 24324866 - 24401655 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0287185
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104200.3 NP_001097670.1 3 - 4 1034 150
GenBank mRNA AB232987.1 - 3 - 4 844 150

View Full Product Details