Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
kl-3
Entrez Gene ID
26067053
Gene Name
male fertility factor kl3
Gene Aliases
CG17629, CG40443, CG40936, CG40992, CG45785, DHC4A, Dmel\CG45785, Dmel_CG45785, Kl-3, Kl3-Y, Lms3, Y104b, kl-4, kl3, ms(Y)104b
Chromosome Location
Chr.Y: 336381 - 563165 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267432
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111012.3 NP_001104482.3 12 - 13 8693 143

View Full Product Details