Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eaf2
Entrez Gene ID
415252
Gene Name
ELL associated factor 2
Gene Aliases
wu:fb76c01, wu:fc43g12, zgc:86600
Chromosome Location
Chr.9: 23003208 - 23007941 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002162.1 NP_001002162.1 3 - 4 437 73
GenBank mRNA BC071305.1 - 3 - 4 437 73

View Full Product Details