Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snx30
Entrez Gene ID
550496
Gene Name
sorting nexin family member 30
Gene Aliases
zgc:112424
Chromosome Location
Chr.10: 1955 - 10533 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017798.1 NP_001017798.1 2 - 3 431 69
GenBank mRNA BC093298.1 - 2 - 3 431 69

View Full Product Details