Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdlim1
Entrez Gene ID
550568
Gene Name
PDZ and LIM domain 1 (elfin)
Gene Aliases
clim1, wu:fa19a03, wu:fy34f10, zgc:110681
Chromosome Location
Chr.13: 25762909 - 25773963 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017870.1 NP_001017870.1 1 - 2 152 120
GenBank mRNA BC092978.1 - 1 - 2 152 120

View Full Product Details