Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
baiap2l1a
Entrez Gene ID
387261
Gene Name
BAR/IMD domain containing adaptor protein 2 like 1a
Gene Aliases
baiap2l1, cb1023, zgc:85624
Chromosome Location
Chr.12: 17815943 - 17863336 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198922.1 NP_944604.1 11 - 12 1345 124
GenBank mRNA BC050238.1 - 11 - 12 1345 124

View Full Product Details