Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
baiap2l1a
Entrez Gene ID
387261
Gene Name
BAR/IMD domain containing adaptor protein 2 like 1a
Gene Aliases
baiap2l1, cb1023, zgc:85624
Chromosome Location
Chr.12: 17815943 - 17863336 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198922.1 NP_944604.1 9 - 10 1065 67
XM_009306593.3 XP_009304868.1 9 - 10 1104 67
GenBank mRNA AY398378.1 - 9 - 10 1066 67
BC050238.1 - 9 - 10 1065 67

View Full Product Details