Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pygmb
Entrez Gene ID
393444
Gene Name
phosphorylase, glycogen, muscle b
Gene Aliases
zgc:63642
Chromosome Location
Chr.7: 22823889 - 22852174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328031.1 NP_001314960.1 19 - 20 2533 123
XM_017356828.2 XP_017212317.1 19 - 20 2374 123
GenBank mRNA BC055181.1 - 8 - 9 952 123

View Full Product Details