Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smpx
Entrez Gene ID
431732
Gene Name
small muscle protein X-linked
Gene Aliases
zgc:91881
Chromosome Location
Chr.24: 25466152 - 25498119 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002185.1 NP_001002185.1 4 - 5 368 111
XM_005162767.4 XP_005162824.1 4 - 5 397 111
XM_009297459.3 XP_009295734.1 4 - 5 364 111
GenBank mRNA BC074039.1 - 4 - 5 368 111

View Full Product Details