Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrpl15
Entrez Gene ID
445041
Gene Name
mitochondrial ribosomal protein L15
Gene Aliases
zgc:92856
Chromosome Location
Chr.7: 58837279 - 58843193 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003435.1 NP_001003435.1 2 - 3 289 107
GenBank mRNA BC076318.1 - 2 - 3 289 107
BC164057.1 - 2 - 3 303 107

View Full Product Details