Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lhx8a
Entrez Gene ID
378959
Gene Name
LIM homeobox 8a
Gene Aliases
cb875, fk78g12, lhx7, lhx8, wu:fk78g12
Chromosome Location
Chr.2: 11205795 - 11210168 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003980.2 NP_001003980.1 5 - 6 778 70
XM_005171154.4 XP_005171211.1 5 - 6 1258 70
GenBank mRNA AY664404.1 - 5 - 6 779 70
BC117601.1 - 5 - 6 766 70

View Full Product Details