Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
extl3
Entrez Gene ID
493783
Gene Name
exostosin-like glycosyltransferase 3
Gene Aliases
im:7141548
Chromosome Location
Chr.20: 49940818 - 50014992 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008402.1 NP_001008402.1 3 - 4 2497 81
XM_005170211.4 XP_005170268.1 4 - 5 3007 81
GenBank mRNA AY786436.1 - 3 - 4 2497 81
BC162406.1 - 3 - 4 2488 81

View Full Product Details