Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
thap1
Entrez Gene ID
692315
Gene Name
THAP domain containing, apoptosis associated protein 1
Gene Aliases
zgc:136597
Chromosome Location
Chr.5: 431958 - 441310 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045284.1 NP_001038749.1 - 296 73
GenBank mRNA BC116602.1 - - 296 73

View Full Product Details