Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taar13e
Entrez Gene ID
100034381
Gene Name
trace amine associated receptor 13e
Gene Aliases
si:dkey-7c18.7, ta66, taar66
Chromosome Location
Chr.20: 46172649 - 46173674 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083043.1 NP_001076512.1 1 - 1 654 139

View Full Product Details