Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
terfa
Entrez Gene ID
192316
Gene Name
telomeric repeat binding factor a
Gene Aliases
CHUNP6903, wu:fi15d05, wu:fi22b03, zgc:111909, zgc:114011
Chromosome Location
Chr.4: 77973874 - 77996958 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173243.2 NP_775350.2 4 - 5 547 57
XM_009301103.3 XP_009299378.1 3 - 4 570 57
XM_009301104.3 XP_009299379.1 3 - 4 571 57
XM_009301105.3 XP_009299380.1 3 - 4 568 57
XM_021474924.1 XP_021330599.1 3 - 4 577 57
XM_021474925.1 XP_021330600.1 3 - 4 575 57
GenBank mRNA AY099522.1 - 7 - 8 749 57
BC098889.1 - 3 - 4 386 57

View Full Product Details