Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
terfa
Entrez Gene ID
192316
Gene Name
telomeric repeat binding factor a
Gene Aliases
CHUNP6903, wu:fi15d05, wu:fi22b03, zgc:111909, zgc:114011
Chromosome Location
Chr.4: 77973874 - 77996958 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173243.2 NP_775350.2 5 - 6 639 64
XM_009301103.3 XP_009299378.1 4 - 5 662 64
XM_009301105.3 XP_009299380.1 4 - 5 660 64
XM_021474924.1 XP_021330599.1 4 - 5 669 64
XM_021474925.1 XP_021330600.1 4 - 5 667 64
GenBank mRNA AY099522.1 - 8 - 9 841 64
BC096857.1 - 4 - 5 463 64
BC098889.1 - 4 - 5 478 64
BC165460.1 - 4 - 5 485 64

View Full Product Details