Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
selenot1a
Entrez Gene ID
352919
Gene Name
selenoprotein T, 1a
Gene Aliases
cb700, selt1a, sept1a, wu:fb63h09, wu:fc79a12
Chromosome Location
Chr.2: 9684305 - 9696323 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178290.6 NP_840075.2 3 - 4 414 105
GenBank mRNA AY221264.1 - 3 - 4 370 105
BC054578.1 - 3 - 4 362 105
CF998986.1 - 3 - 4 379 105
CK681469.1 - 3 - 4 393 105
CK691600.1 - 3 - 4 392 105
CK699389.1 - 3 - 4 392 105
DQ660904.1 - 3 - 4 374 105
EH438162.1 - 3 - 4 412 105

View Full Product Details